Toallitas para salas blancas, presaturadas y estériles, AlphaSat® 10
Proveedor: Texwipe
TX3280
115-0488EA 995 EUR
115-0488
Toallitas para salas blancas, presaturadas y estériles, AlphaSat® 10
Toallitas Toallitas para salas limpias Presaturadas
These 100% continuous filament polyester wipe with thermally sealed borders have been designed to control particle and fibre contamination. They are pre-wetted with 70% USP Isopropyl Alcohol (IPA) and 30% Water for Injection (WFI).
  • Ideal for wiping gloved hands
  • Optimises solution usage and eliminates overspray of alcohol
  • Reduces IPA spill potential and eliminates the storage and disposal of excess solvents
  • Superior wipe strength allows for rigorous wiping with minimal release of fibres and particles
Ideal for cleaning critical sterile environments inMore Product Information