VWR® B´Mat, Permanent cleanroom mats
Proveedor: VWR Collection
115-1832EA 0 EUR
115-1832 115-1831 115-1807 115-1833 115-1834 115-1835 115-1836 115-1808 115-1801 115-1802 115-1837 115-1838 115-1839 115-1840 115-1842 115-1841 115-1847 BASABMAT080X120CM BASABMAT100X100CM BASABMAT120X250CM BASABMAT090X250CM BASABMAT090X300CM BASABMAT040X120CM BASABMAT120X135CM BASABMAT120X180CM BASABMAT120X210CM BASABMAT085X140CM BASABMAT080X150CM BASABMAT080X170CM BASABMAT100X230CM BASABMAT130X400CM BASABMAT230X250CM
VWR® B´Mat, Permanent cleanroom mats
Alfombrillas Alfombras de suelo permanentes
Permanent mats are installed to provide the highest level of cleanliness. The life of mats strongly depends on the volume, and degree of cleanliness of the foot and wheeled traffic. Solvent-based cleaning materials should, on no account, be used on these mats: They will adversely affect both the function and appearance. To avoid risk of slipping, it is also important that both the mats and the flooring should be completely dry.
  • Special polymer (bacteriostatic, ESD, non outgassing)
  • Attracts and retains over 98% of foot- and wheel-borne particles
  • Bevelled