Swabs, with cotton heads
Proveedor: Texwipe
STX705P STX705W
115-1881EA 57.8 EUR
115-1881 115-1882
Swabs, with cotton heads
Hisopos Hisopos para limpieza
Sterile cotton swabs are constructed from 100% USP grade cotton. Bonded securely to a shaft by an aqueous-based adhesive. Made to exacting and consistent tolerances using high precision automated processes.
  • Gamma irradiated pre-sterilised swabs eliminating repackaging, documentation and validation of autoclaved swabs
  • Lot number and expiration date on each package and in barcode for inventory controls
  • Certificates of compliance to confirm irradiation dose with specifications
  • Clear packaging which offers visibility of seal integrity
Envase: Easy to open,More Product Information