Dual-wire fabric wristband, SCS
Proveedor: DESCO
2360 2368 2371
115-3057EA 33.4 EUR
115-3057 115-3058 115-3059
Dual-wire fabric wristband, SCS
Tomas de tierra Muñequeras antiestáticas
The Dual-wire fabric wristband is available in one size that adjusts to almost any size wrist for reliable protection.
  • Adjustable, elastic nylon material with conductive woven thread
  • Fits most wrist sizes to provide reliable protection
  • Dual-Wire conductor provides redundancy
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
The wristband provides full conductivity, comfort, and reliability, while ensuring continuous drain of static charge when used with SCS 2300 Dual-wire coil cords (soldMore Product Information