Signs, floor signs, ToughStripe™, divider 'T' marks
Proveedor: Brady
104451
116-0923EA 64.8 EUR
116-0923
Signs, floor signs, ToughStripe™, divider 'T' marks
Señales
ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.
  • Floor marking products are made with B-514 polyester
  • Recommended for indoor use and dry environment
  • Resistant to harsh chemicals
0,203 mm thickness and service temperature range of ⁻18 °C to 54 °C.
Certificaciones: English XL Variable Incidence Tribometer per ASTM / ANSI F1679. ANSI A1264.2­2006 and OSHA 1910.22