Lithium tetraborate : lithium metaborate (66.5:33.5) % (w/w), Spectroflux® 110
Proveedor: Thermo Fisher Scientific
12086.A1 12086.D9
12086.A1EA 0 EUR
12086.A1 12086.D9
Lithium tetraborate : lithium metaborate (66.5:33.5) % (w/w), Spectroflux® 110
Lithium tetraborate : lithium metaborate
Intermediate activity flux. Suitable for aluminosilicates and calcareous refractories. Often used in testing of cement and steel. Samples must be fully oxidized
Spectroflux 110, Lithium tetraborate & Lithium metaborate, 66.5:33.5 w/w% is used to dissolve aluminosilicates ranging from 100% silicon dioxide to 100% aluminum(III) oxide. It has intermediate acidity and suitable for aluminosilicates and calcareous refractories.