Cobre (II) nitrato 2,5-hidrato 98.0-102.0% ACS

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate

12523.36 12523.18 12523.A3 12523.A9
12523.36EA 99.5 EUR
12523.36 12523.18 12523.A3 12523.A9
Cobre (II) nitrato 2,5-hidrato 98.0-102.0% ACS
Cobre (II) nitrato 2,5-hidrato

Purity: 98.0-102.0% CAS No.: 19004-19-4 Molecular Formula: CuN2O6.2.5H2O Formula Weight: 232.59 (187.55anhy) Physical Form: Crystalline Lumps Density: 2.32 Appearance: Blue Odour: Odourless Melting Point: ca 115° Boiling Point: 170° dec. Solubility: Soluble in water and alcohol Merck Reference: 14,2643 UN No.: 3085 MDL No.: MFCD00149671


Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, Colouring copper black, and producing a burnished effect on iron

Formula: Cu(NO₃)₂·2.5H₂O
MW: 232.59 g/mol
Pto. Ebullición: 170 °C (desc.)
Pto. de fusión: ∼115 °C
Densidad: 2.32
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02253001
Núm. CAS: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1 III
Índice Merck: 14,2643

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR