Selenato de sodio, anhidro ≥99.8% (metals basis)

Proveedor: Thermo Fisher Scientific
Peligro

12613.18 12613.06 12613.30 12613.A1
12613.18EA 74.3 EUR
12613.18 12613.06 12613.30 12613.A1
Selenato de sodio, anhidro ≥99.8% (metals basis)
Selenato de sodio

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.


Stable under alkaline and oxidizing conditions.


As a reagent

Formula: Na₂O₄Se
Pto. de fusión: 32 °C trans,
Densidad: 3,213
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003490
Núm. CAS: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Índice Merck: 14,8671

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR