VWR®, Biohazard Bags, PP
Proveedor: VWR Collection
129-0024EA 140 EUR
129-0024 129-0025 129-0027 129-1564
VWR®, Biohazard Bags, PP
Bolsas Bolsas de eliminación de residuos
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Complete with wire closures that allow expansion of the bag and ventilation when cooling. Highly resistant to tears and punctures. Available as transparent bags without temperature indicator, or orange with temperature indicator. PP, 50 µm thick, with 'Biohazard' label.
  • Autoclavables hasta 135 °C
  • El texto del indicador de temperatura es blanco antes del autoclavado y negro después
  • 'Contents Have Been Autoclaved' indica que la esterilización en autoclave se ha