Cleaning mitt wipes, PolyMitt™
Proveedor: MICRONOVA
MW-611 MWD-611 MW-AA MW-6 MW-2011 MW-20 MW-11 MW-6IR MWDT-611
112-1293EA 17.8 EUR
112-1293 129-0618 129-0617 129-0616 129-0663 129-0615 MICRMW-11 MICRMW-6IR MICRMWDT-611
Cleaning mitt wipes, PolyMitt™
Toallitas Paños de limpieza reutilizables Paños para tareas delicadas
These cleaning mitts offer the versatility of a high quality wipe with the convenience and security of a glove. Designed for simplified cleaning around hard to reach areas in the cleanroom.
  • Durable cleaning mitts for simplified cleaning
  • Versatility of a high quality wipe with the convenience and security of a glove
  • Dimensions: 300×200 mm
  • For aseptic applications: Available irradiated or can be autoclaved prior to use
  • Available with or without a urethane foam layer
  • Compatible with strong disinfectants
  • Reusable and autoclavable
The Micronova PolyMitt™More Product Information