Autoclavable Bags, Dispenser Box
Proveedor: RATIOLAB
7001000 7001005 7001401 7001405
129-9811EA 12.4 EUR
129-9811 RATI7001005 129-0703 RATI7001405
Autoclavable Bags, Dispenser Box
Bolsas Bolsas de eliminación de residuos
Bags of very strong PP-film with a broad seam at the bottom, for the disposal of contaminated labware.
  • Volume approx. 1.5 L, film thickness 50 µm
  • For bag size 200×300 mm: Practical bench-top stand, plastic-coated, with 3 rubber feet
For autoclaving, the bag must not be completely closed. Standard bags are autoclavable (20 min.) at 134 °C, high transparency bags at 121 °C.