VWR®, Autoclavables bolsas
Proveedor: VWR Collection
129-0018EA 464 EUR
129-0018 129-0019 129-1153 129-1154 129-1155 129-1156 129-1570
VWR®, Autoclavables bolsas
Bolsas Bolsas de eliminación de residuos
Sturdy, temperature resistant bags, ideal for disposing of hazardous waste that must first be autoclaved. Can also be used for autoclaving other products. Supplied complete with sealing wires. PP, 37,5 µm or 50 µm thick, transparent.
  • Claro bolsas
  • Las bolsas son autoclavables hasta 135 °C
  • Gran resistencia a los desgarros e impactos