Sterilisation flat reels, Stericlin®
Proveedor: VP Medical Packaging
3FKFS230114 3FKFS230120 3FKFS230106 3FKFS230110 3FKFS230102 3FKFS230104 3FKFS230108 3FKFS230112 3FKFS230116 3FKFS230118
129-1278EA 212 EUR
129-1278 129-1281 129-1274 129-1276 129-1272 129-1273 129-1275 129-1277 129-1279 129-1280
Sterilisation flat reels, Stericlin®
Bolsas Bolsas de esterilización
These see-through sterilisation reels made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certificaciones: DIN EN ISO 11607 and DIN 868-5.