Autoclavable Peel Pouches, with Steam Indicator, Self Seal, 1073B Tyvek® / Polyester
Proveedor: KEYSTONE CLEANROOM PRODUCTS
BBMGTVKSS0206 BBMGTVKS02095 BBMGTVKSS0306 BBMGTVKSS0308 BBMGTVKS03508 BBMGTVKSS0415 BBMGTVKSS0422 BBMGTVKSS0510 BBMGTVKSS0608 BBMGTVKSS0612 BBMGTVKSS0628 BBMGTVKSS0632 BBMGTVKSS0635 BBMGTVKSS0713 BBMGTVKSS0810 BBMGTVKSS0815 BBMGTVKSS0818 BBMGTVKSS0834 BBMGTVKSS1012 BBMGTVKSS1020 BBMGTVKSS1214 BBMGTVKSS1217 BBMGTVKSS1424 BBMGTVKSS1520 BBMGTVKSS1617 BBMGTVKSS1626 BBMGTVKSS1632 BBMGTVKSS1635 BBMGTVKSS1822 BBMGTVKSS2426 BBMGTVKSS2430 BBMGTVKSS2435 BBMGTVKS163525
129-1657EA 1700 EUR
129-1657 129-2471 129-1658 129-1659 129-2472 129-1660 129-1661 129-1662 129-1663 129-1664 129-1665 129-1666 129-1667 129-1668 129-1669 129-1670 129-1671 129-1672 129-1673 129-1674 129-1675 129-1676 129-1677 129-1678 129-1679 129-1680 129-1681 129-1682 129-1683 129-1684 129-1685 129-1686 129-2473
Autoclavable Peel Pouches, with Steam Indicator, Self Seal, 1073B Tyvek® / Polyester
Bolsas Bolsas de esterilización
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packed in a controlled cleanroom environment
  • Easy, fast and reliable self-sealing closure system
  • Integrated steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • One side made of hydrophobic and lint-free Tyvek® 1073B
  • One side made of laminated polyester/polyolefin film
  • Peels open cleanly
A significant upgrade over paper-based steam sterilisation pouches. These Tyvek® peel pouches will not rip orMore Product Information