Autoclave Self-Seal Bags with Steam Indicator, Tyvek® 1073B
Proveedor: KEYSTONE CLEANROOM PRODUCTS
COVMGTVK1X14SS COVMGTVK3X6SS COVMGTVK3X36SS COVMGTVK5X25SS COVMGTVK6X8SS COVMGTVK6X16SS COVMGTVK6X22SS COVMGTVK6X34SS COVMGTVK8X9SS COVMGTVK8X12SS COVMGTVK8X14SS COVMGTVK8X30SS COVMGTV10X12SS COVMGTV10X20SS COVMGTV10X25SS COVMGTV12X12SS COVMGTV12X16SS COVMGTV12X18SS COVMGTV12X24SS COVMGTV12X32SS COVMGTV12X34SS COVMGTV14X22SS COVMGTV14X28SS COVMGTV14X42SS COVMGTV15X18SS COVMGTV16X24SS COVMGTV16X36SS COVMGTV16X44SS COVMGTV18X22SS COVMGTV18X24SS COVMGTV18X26SS COVMGTV18X30SS COVMGTV18X40SS COVMGTV18X68SS COVMGTV20X20SS COVMGTV20X32SS COVMGTV23X30SS COVMGTV24X24SS COVMGTV24X44SS COVMGTV25X32SS COVMGTV25X37SS COVMGTV26X28SS COVMGTV26X30SS COVMGTV30X30SS COVMGTV30X22SS COVMGTV30X32SS COVMGTV32X34SS COVMGTV34X34SS COVMGTV34X40SS COVMGTV36X36SS COVMGTV36X42SS COVMGTV36X48SS COVMGTV36X60SS COVMGTV38X40SS COVMGTV38X48SS COVMGTV40X48SS COVMGTV42X46SS COVMGTV48X28SS COVMGTV48X48SS COVMGTV48X64SS COVMGTVKUB4X23 COVMGTVK1626SS COVMGTVK4X16SS COVMGTVK4230SS COVMGTVK4X23SS COVMGTVK416SS COVMGTVKUB1218
129-2661EA 810 EUR
129-2661 129-2670 129-2669 129-2679 129-2733 129-2680 129-2681 129-2732 129-2737 129-2734 129-2735 129-2736 129-2834 129-2835 129-2836 129-2837 129-2838 129-2839 129-2840 129-2841 129-2842 129-2843 129-2844 129-2845 129-2846 129-2847 129-2848 129-2849 129-2850 129-2851 129-2852 129-2853 129-2854 129-2855 129-2856 129-2857 129-2858 129-2859 129-2860 129-2861 129-2862 129-2863 129-2864 129-2866 129-2865 129-2760 129-2683 129-2684 129-2685 129-2686 129-2687 129-2688 129-2689 129-2690 129-2691 129-2692 129-2693 129-2694 129-2695 129-2696 129-2741 129-2710 129-2677 129-2672 129-2678 KACCCOVMGTVK416SS KACCCOVMGTVKUB1218
Autoclave Self-Seal Bags with Steam Indicator, Tyvek® 1073B
Bolsas Bolsas de esterilización
Self-sealing Tyvek® 1073B bags are ideal for steam sterilisation of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges. No need to use or validate a heat sealing machine.
  • Manufactured and packed in a controlled cleanroom environment
  • Convenient self-sealing closure system
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic and lint-free
  • 100% medical grade Tyvek® 1073B allows fast steam penetration
  • Excellent impact resistance with