Elasticised Covers for Steam Sterilisation, Rectangular Shaped, DuPont™ Tyvek® 1422A
Proveedor: KEYSTONE CLEANROOM PRODUCTS
T2ASHEET0324E T2ASHEET1818E T2ASHEET2424E T2ASHEET2440E T2ASHEET2626E T2ASHEET2816E T2ASHT2838E2D2 T2ASHEET3014E T2ASHEET3024E T2ASHEET3434E T2ASHEET4080E T2ASHEET4233E T2ASHEET4320E T2ASHEET2436E T2ASHEET3034E
129-3040EA 3360 EUR
129-3040 129-3054 129-3013 129-2976 129-2977 129-2978 129-2995 129-2979 129-2980 129-2983 129-2987 129-2988 129-2989 129-2975 129-2982
Elasticised Covers for Steam Sterilisation, Rectangular Shaped, DuPont™ Tyvek® 1422A
Cubiertas
Save time and standardise the activities in equipment preparation for sterilisation. Leave the covers in place during installation to prevent microbial and particulate contamination. Leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam Sterilisation Indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Elasticised Tyvek® Covers provide a convenient, time-More Product Information