Gamma irradiated cleanroom pens, BIC®
Proveedor: KEYSTONE CLEANROOM PRODUCTS
S01PENBLK S01PENBLUE S02PENBLK S02PENBLUE S05PENBLK S05PENBLUE S10PENBLK S10PENBLUE
129-3107EA 1770 EUR
129-3107 129-3108 129-3117 129-3118 129-2940 129-2941 129-2946 129-2947
Gamma irradiated cleanroom pens, BIC®
Bolígrafos Bolígrafos para salas limpias
These pens and markers eliminate the risks associated with surface sanitisation procedures.
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek® peel pouches
  • Minimise contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing intruments
CRP offers gamma irradiated pens and markers for the convenience of pharmaceutical cleanroom operations. These are a 'Ready to Use Option' when sterilisation on-site is not possible; or surface sterilisation is insufficient. The clean Tyvek® peel pouch packagingMore Product Information