Autoclave Bags with Steam Indicator and Drawstring, DuPont™ Tyvek® 1422A
Proveedor: KEYSTONE CLEANROOM PRODUCTS
BGT2A0418D BGT2A0425D BGT2A0426D BGT2A0428D BGT2A0505D18 BGT2A0506D BGT2A0508D BGT2A0509D BGT2A0510D BGT2A0511D BGT2A0606D BGT2A0608D BGT2A0609D BGT2A0610D BGT2A0615D BGT2A0616D BGT2A0636D BGT2A0707D BGT2A0709D BGT2A0710D BGT2A0713D BGT2A0811D BGT2A0812D BGT2A0815D BGT2A0822D BGT2A09512D BGT2A1012D BGT2A1018D BGT2A1020D BGT2A1024D BGT2A1030D BGT2A1064D BGT2A1072D BGT2A1212D BGT2A1214D BGT2A1216D BGT2A1218D BGT2A1220D BGT2A1222D BGT2A1224D BGT2A1228D BGT2A1230D BGT2A1232D BGT2A1236D BGT2A1242D BGT2A1414D BGT2A1420D BGT2A1422D BGT2A1424D BGT2A1426D BGT2A1436D BGT2A1438D BGT2A1440D BGT2A1448D BGT2A1526D BGT2A1543D BGT2A1616D BGT2A1620D BGT2A1624D BGT2A1630D BGT2A1632D BGT2A1636D BGT2A1638D BGT2A1644D BGT2A1648D BGT2A1820D BGT2A1822D BGT2A1824D BGT2A1830D BGT2A1832D BGT2A1836D BGT2A1840D BGT2A1846D BGT2A1848D BGT2A1854D BGT2A1870D BGT2A2020D BGT2A2024D BGT2A2030D BGT2A2032D BGT2A2036D BGT2A2040D BGT2A2044D BGT2A2046D BGT2A2048D BGT2A2424D BGT2A2436D BGT2A2440D BGT2A2448D BGT2A2634D BGT2A2646D BGT2A2828D BGT2A2832D BGT2A3028D BGT2A3030D BGT2A3232D BGT2A3248D BGT2A3434D BGT2A3628D BGT2A3636D BGT2A3642D BGT2A3646D BGT2A3648D BGT2A3664D BGT2A3672D BGT2A3838D BGT2A4040D BGT2A4060D BGT2A4244D BGT2A4450D BGT2A4848D BGT2A4850D BGT2A4864D BGT2A5028D BGT2A6250D BGT2A0518D BGT2A0604D BGT2A0910D BGT2A0917D BGT2A3220D BGT2A3638D BGT2A1038D BGT2A4580D BGT2A1240D BGT2A2648D BGT2A3436D
129-3157EA 2800 EUR
129-3157 129-3156 129-3154 129-3158 129-3172 129-3174 129-3175 129-3176 129-3177 129-3178 129-3185 129-3187 129-3259 129-3261 129-3265 129-3267 129-3269 129-3273 129-3275 129-3278 129-3279 129-3283 129-3286 129-3287 129-3225 129-3233 129-3234 129-3237 129-3240 129-3244 129-3247 129-3254 129-3255 129-3257 129-3591 129-3594 129-3597 129-3599 129-3604 129-3610 129-3612 129-3614 129-3617 129-3622 129-3627 129-3555 129-3564 129-3571 129-3573 129-3576 129-3578 129-3580 129-3582 129-3584 129-3587 129-3665 129-3671 129-3674 129-3677 129-3679 129-3691 129-3693 129-3696 129-3690 129-3688 129-3685 129-3683 129-3630 129-3633 129-3635 129-3638 129-3640 129-3642 129-3644 129-3646 129-3649 129-3652 129-3655 129-3660 129-3662 129-3432 129-3435 129-3438 129-3440 129-3442 129-3451 129-3456 129-3459 129-3463 129-3404 129-3406 129-3410 129-3412 129-3425 129-3428 129-3504 129-3507 129-3514 129-3520 129-3525 129-3530 129-3534 129-3537 129-3545 129-3467 129-3469 129-3473 129-3476 129-3481 129-3485 129-3490 129-3492 129-3496 129-3499 129-2799 129-3180 129-3183 129-3230 129-3231 129-3503 129-3527 129-3249 129-3487 129-3625 129-3408 129-3516
Autoclave Bags with Steam Indicator and Drawstring, DuPont™ Tyvek® 1422A
Bolsas Bolsas de esterilización
Tyvek® 1422A autoclave bags with drawstrings can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Tear and puncture resistant
  • Fast and consistent for equipment preparation area
  • Steam Indicator
  • Hydrophobic
  • Lint-free
  • Strong factory seals
  • Exceptional microbial barrier
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use of these drawstring bags will also save valuable time in the equipment prep area.
The drapeable nature of the Tyvek® 1422A material is ideal forMore Product Information