Steam sterilisation chevron peel pouch with steam indicator and hang hole, isolator bags, self-sealing, Tyvek® 1073B/PE
Proveedor: KEYSTONE CLEANROOM PRODUCTS
BBMGTVKS0206HH BBMGTVK02095HH BBMGTVKS0306HH BBMGTVKS0308HH BBMGTV03508HH BBMGTVKS0415HH BBMGTVKS0422HH BBMGTVKS0510HH BBMGTVKS0608HH BBMGTVKS0612HH BBMGTVKS0628HH BBMGTVKS0632HH BBMGTVKS0635HH BBMGTVKS0713HH BBMGTVKS0810HH BBMGTVK0815HH BBMGTVKS0818HH BBMGTVKS0834HH BBMGTVKS1012HH BBMGTVK1020HH BBMGTVKS1214HH BBMGTVKS1217HH BBMGTVKS1424HH BBMGTVKS1520HH BBMGTVKS1617HH BBMGTVKS1626HH BBMGTVKS1632HH BBMGTVK1635HH BBMGTVKS1822HH BBMGTVKS2426HH BBMGTVKS2430HH BBMGTVKS2435HH
129-3304EA 1960 EUR
129-3304 129-3350 129-3305 129-3306 129-3349 129-3307 129-3308 129-3309 129-3310 129-3313 129-3314 129-3315 129-3316 129-3317 129-3318 129-3351 129-3319 129-3320 129-3321 129-3352 129-3322 129-3323 129-3324 129-3325 129-3390 129-3391 129-3392 129-3353 129-3393 129-3394 129-3395 129-3396
Steam sterilisation chevron peel pouch with steam indicator and hang hole, isolator bags, self-sealing, Tyvek® 1073B/PE
Bolsas Bolsas de esterilización
Improve the prep time, performance, microbial barrier, and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Integrated Hang Hole is ideal for use with Isolator VHP decontamination cycles
  • Easy, fast, and reliable self-sealing closure system
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly and lint-free
  • Chevron peel and hydrophobic
  • Tear and puncture resistant
  • Integrated Steam Sterilisation Indicator
A significant upgrade over paper-based steam sterilisation pouches. Integrated hang-hole for useMore Product Information