Gamma Irradiated Sterility Maintenance Covers, Self-Seal
Proveedor: KEYSTONE CLEANROOM PRODUCTS
3312ST 3315ST 3315ST5 3320ST 3322ST 3330ST 3334ST 3335ST 3320ST3 3322ST3 3334ST3 3334STJ
129-3337EA 3560 EUR
129-3337 129-3338 129-3339 129-3340 129-3341 129-3342 129-3343 129-3344 KACC3320ST3 KACC3322ST3 KACC3334ST3 KACC3334STJ
Gamma Irradiated Sterility Maintenance Covers, Self-Seal
Bolsas Bolsas de esterilización
Ideal for pharmaceutical and biotech production applications.
  • Wide range of sizes
  • Convenient self-seal closure; no need for the use of or validation of a heat sealer
  • No tools required to open
  • Impervious to liquid sanitisation agents
  • Visibility inside the bag allows for identification
  • Prolong the storage and shelf-life of products sterilized ‘in-house’
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilized items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protectsMore Product Information