Autoclave Bags with Steam Indicator, Self-Seal, Heavy Duty SMS
Proveedor: KEYSTONE CLEANROOM PRODUCTS
BGSMS3SS09.516 BGSMS3SS1217 BGSMS3SS1221 BGSMS3SS1318 BGSMS3SS14.522 BGSMS3SS1520 BGSMS3SS1622 BGSMS3SS1824 BGSMS3SS1830 BGSMS3SS2028 BGSMS3SS2126 BGSMS3SS2424 BGSMS3SS2430 BGSMS3SS2435 BGSMS3SS2630 BGSMS3SS2836 BGSMS3SS3030 BGSMS3SS3636 BGSMS3SS3648 BGSMS3SS4848 BGSMS3SS1448 BGSMS3SS7550
KACCBGSMS3SS09.516EA 1540 EUR
KACCBGSMS3SS09.516 KACCBGSMS3SS1217 KACCBGSMS3SS1221 KACCBGSMS3SS1318 KACCBGSMS3SS14.522 KACCBGSMS3SS1520 KACCBGSMS3SS1622 KACCBGSMS3SS1824 KACCBGSMS3SS1830 KACCBGSMS3SS2028 KACCBGSMS3SS2126 KACCBGSMS3SS2424 KACCBGSMS3SS2430 KACCBGSMS3SS2435 KACCBGSMS3SS2630 KACCBGSMS3SS2836 KACCBGSMS3SS3030 KACCBGSMS3SS3636 KACCBGSMS3SS3648 KACCBGSMS3SS4848 129-4025 KACCBGSMS3SS7550
Autoclave Bags with Steam Indicator, Self-Seal, Heavy Duty SMS
Bolsas Bolsas de esterilización
Self Sealing Heavy Duty SMS Autoclave Bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilisation of critical pharmaceutical equipment and components.
  • Convenient, time saving self-seal closure system
  • Significant improvement versus cutting and wrapping with sheets and tape
  • Superior barrier prevents liquid and microbial penetration
  • Manufactured and packaged in a certified ISO 6 Cleanroom
  • Strong, rip and tear resistant 50 gsm SMS material
  • Sub packed into sealed labelled poly bags with manufacturing lot