Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®

Proveedor: Thermo Fisher Scientific
Atención

13029.14 13029.22
13029.14EA 167 EUR
13029.14 13029.22
Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®
Bismuth germanium oxide

Purity: 99.9995% CAS No.: 12233-56-6 Molecular Formula: Bi4Ge3O12 Formula Weight: 1245.68 Physical Form: Powder Appearance: White Odour: Odourless MDL No.: MFCD00064629

Formula: Bi₄Ge₃O₁₂
MW: 1245.68 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064629
Núm. CAS: 12233-56-6
EINECS: 235-457-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR