Imiquimod Related Compounds, Reference Standard
Proveedor: USP
1338324 1338335 1338346 1338357
1338324.EA 1230 EUR
1338324. 1338335. 1338346. 1338357.
Imiquimod Related Compounds, Reference Standard
Normativas Estándares orgánicos
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificaciones: US Pharmacopea Reference Standard