Sodio hidrogenocarbonato 99.0-100.5%, en polvo, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E500, Fluka™

Proveedor: Honeywell Chemicals

13433-25KG 13433-1KG 13433-5KG 13433-6X1KG
13433-25KGEA 163 EUR
13433-25KG 13433-1KG 13433-5KG 13433-6X1KG
Sodio hidrogenocarbonato 99.0-100.5%, en polvo, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E500, Fluka™
Sodio hidrogenocarbonato
Formula: NaHCO₃
Pto. de fusión: Dec.
Densidad: 2,2 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003528
Núm. CAS: 144-55-8
EINECS: 205-633-8
Índice Merck: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay 99.0 - 100.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Water insoluble matter complying
pH (5 %, 20°C) Max. 8.6
Loss on drying (via silica gel) Max. 0.2 %
Aluminium (Al) Max. 0.5 ppm
Ammonium (NH₄) Max. 20 ppm
Ammonia (NH₃) complying
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 100 ppm
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.01 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 5 ppm
Carbonate (CO₃) complying
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
S-compounds (as SO₄) Max. 150 ppm
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR