Neon 2 Production and Laboratory Chairs, with Synchronous Technology
Proveedor: bimos
9573-SP02-3280 9573-MG01-3278 9573-2000-3278 9573-2000-3279 9573-2000-3280 9573-2001-3278 9573-2001-3280 9573-MG01-3279 9573-MG01-3280 9573-MG02-3278 9573-MG02-3279 9573-MG02-3280 9573-SP01-3278 9573-SP01-3279 9573-SP01-3280 9573-SP02-3278 9573-SP02-3279 9573-2001-3279 9573-MG11-3278 9573-2002-3278 9573-2002-3280 9573-MG11-3279 9573-MG11-3280 9573-SP11-3278 9573-SP11-3279 9573-SP11-3280 9573-2002-3279 9573-6802-3278 9573-6801-3278 9573-6801-3280 9573-6802-3279 9573-6802-3280 9573-6811-3278 9573-6811-3279 9573-6811-3280 9573-6801-3279 9573EMG0132787 9573EMG01-3279 9573-6801-3277 9573MG01328052 9573-MG01-3277 9573-2000-3277 9573-SP01-3277 9573-SP11-3277 9575-MG01-3280 9576EPU277709 9573EMG0127852 9573-MG02-3277
139-0693EA 689 EUR
139-0693 139-0679 139-0661 139-0662 139-0663 139-0664 139-0666 139-0680 139-0681 139-0682 139-0683 139-0684 139-0688 139-0689 139-0690 139-0691 139-0692 139-0665 139-0685 139-0667 139-0669 139-0686 139-0687 139-0694 139-0695 139-0696 139-0668 139-0673 139-0670 139-0672 139-0674 139-0675 139-0676 139-0677 139-0678 139-0671 BIMO9573EMG0132787 BIMO9573EMG01-3279 BIMO9573-6801-3277 BIMO9573MG01328052 BIMO9573-MG01-3277 BIMO9573-2000-3277 BIMO9573-SP01-3277 BIMO9573-SP11-3277 BIMO9575-MG01-3280 BIMO9576EPU277709 BIMO9573EMG0127852 BIMO9573-MG02-3277
Neon 2 Production and Laboratory Chairs, with Synchronous Technology
Sillas
Neon supports the forward-leaning sitting position that is often necessary in lab work. Its modern design matches the high-tech laboratory environment perfectly and of course it has all the characteristics you would expect of a good laboratory chair. It is seamless, easy to clean, washable, and resistant to disinfectants. So, Neon is the ideal solution for the laboratory, when a really comfortable ergonomic chair is needed.
  • With castors
  • With synchronous technology
  • Ergonomic package with height, seat inclination, seat depth and backrest height adjustment,