Triisopropanolato de aluminio ≥98%

Proveedor: Thermo Fisher Scientific
Peligro

14007.A1 14007.A7
14007.A1EA 65.9 EUR
14007.A1 14007.A7
Triisopropanolato de aluminio ≥98%
Triisopropanolato de aluminio

Purity: 98+% CAS No.: 555-31-7 Molecular Formula: Al[OCH(CH3)2]3 Formula Weight: 204.25 Physical Form: Granular Density: 1.035 Refractive Index: 1.0346 Sensitivity: Moisture Sensitive Melting Point: 131-135° Boiling Point: 135°/10mm Solubility: Soluble in alcohol, benzene, and toluene. Decomposes in water Merck Reference: 14,347 UN No.: 3181 MDL No.: MFCD00008870


Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates

Formula: C₉H₂₁AlO₃
Pto. Ebullición: 135 °C (10 mmHg)
Pto. de fusión: 131…135 °C
Densidad: 1,035
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008870
Núm. CAS: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Índice Merck: 14,347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR