Disinfectants for use in Ultrasonic baths, STAMMOPUR 24
Proveedor: BANDELIN-ELECTRONIC
977 978
142-2012EA 102 EUR
142-2012 142-2013
Disinfectants for use in Ultrasonic baths, STAMMOPUR 24
Soluciones de limpieza Desinfectantes
STAMMOPUR 24 is formulated for cleaning and disinfection of respirators, protective equipment, medical tnstruments and laboratory equipment. The product was especially developed for cleaning and disinfection in an ultrasonic bath and allows short sonication times at low dosage.
  • Rapid disinfection and efficient cleaning in ultrasonic baths
  • Broad range spectrum of activity- bactericidal, tuberculocidal, yeasticidal
  • Virucidal against BVDV, Vaccinia, HBV, HCV, HIV, A-H5N1
  • High material compatibility: Metal, glass, porcelain, plastics and rubber