Triacetato de tulio hidrato ≥99.9% (REO, rare earth oxide basis), REacton®

Proveedor: Thermo Fisher Scientific
Atención

14582.04 14582.09
14582.04EA 152 EUR
14582.04 14582.09
Triacetato de tulio hidrato ≥99.9% (REO, rare earth oxide basis), REacton®
Triacetato de tulio hidrato

Purity: 99.9% CAS No.: 314041-04-8 Molecular Formula: Tm(OOCCH3)3.xH2O Formula Weight: 346.07(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00013046

Formula: C₆H₉O₆Tm·H₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00013046
Núm. CAS: 314041-04-8
EINECS: 254-326-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR