Gas detection tubes for hydrochloric acid
Proveedor: Dräger
CH29501 6728181 8103481
151-0203EA 105 EUR
151-0203 151-0152 118-0328
Gas detection tubes for hydrochloric acid
Tubos per gas
To measure the 'spot concentrations' of hydrochloric acid in the air with an Accuro® pump - Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
  • Hydrogen sulphide and sulphur dioxide do not interfere in the TLV range
  • Impossible to measure hydrochloric acid in the presence of other mineral acids
  • Chlorine and nitrogen dioxide are indicated but with different sensitivities
One stroke isMore Product Information