Bis(trimethylsilyl)acetamide + Trimethylchlorosilane, (BSA+TMCS) BSA 93 - 97%, LiChropur™ for GC derivatisation, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

15256-10ML
15256-10MLEA 62.9 EUR
15256-10ML
Bis(trimethylsilyl)acetamide + Trimethylchlorosilane, (BSA+TMCS) BSA 93 - 97%, LiChropur™ for GC derivatisation, Supelco®
Bis(trimethylsilyl)acetamide + Trimethylchlorosilane, (BSA+TMCS)

Suitable for the derivatization of a-ketoacids, amines, amino acids, cycloserine, histamine and metabolites, phenylethylamines, phosphatidylserines and tryptamines.


Trimethylchlorosilane TMCS (minor component): 3.0 - 5.0% (GC)


BSA (major component): 93,0 - 97,0% (GC)

Temperatura de almacenaje: Nevera
UN: 2924
ADR: 8(3) II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR