Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Proveedor: Cytiva
17-5498-51 17-5498-52 17-5498-53 17-5498-54
17-5498-51EA 227 EUR
17-5498-51 17-5498-52 17-5498-53 17-5498-54
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Columnas de cromatografía
These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.
  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH
  • Convenient HiTrap™
More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Capacidad de adsorción ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
~ 80 mg polyclonal IgG/ml resin, 6 min residence time
Presión máx. 5 bar
Rango de pH 3 – 12
Temperature limit 40 °C
Almacenamiento 20% ethanol, 2 to 8 °C