Immobiline™ DryStrip gels
Proveedor: Cytiva
17-6001-13 17-6001-15 17-6001-95 17-6001-96 17-6002-44 17-6002-46 17-6002-47 17-1233-01 17-1234-01 17-1235-01 17-6001-10 17-6001-11 17-6001-12 17-6001-14 17-6001-94 17-6001-97 17-6002-45 18-1016-60 18-1016-61 17-6003-73 17-6003-74 17-6003-75 17-6003-76 17-6003-77
17-6001-13EA 191 EUR
17-6001-13 17-6001-15 17-6001-95 17-6001-96 17-6002-44 17-6002-46 17-6002-47 17-1233-01 17-1234-01 17-1235-01 17-6001-10 17-6001-11 17-6001-12 17-6001-14 17-6001-94 17-6001-97 17-6002-45 18-1016-60 18-1016-61 17-6003-73 17-6003-74 17-6003-75 17-6003-76 17-6003-77
Immobiline™ DryStrip gels
Geles de electroforesis Geles prefabricados
Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. IPG strips contain a preformed pH gradient immobilised in precast polyacrylamide gels on a plastic backing. A comprehensive range of overlapping IPG strips covering narrow, medium, and wide pH ranges are available in various strip lengths, and the strips are individually packaged and bar coded for convenient handling and sample tracking.
  • Improved Immobiline™ DryStrip gels provide increased stability, reproducibility, and protein