Tris(trimethylsilyl)phosphite ≥96%

Proveedor: Thermo Fisher Scientific
Atención

17159.02 17159.04
17159.02EA 34.1 EUR
17159.02 17159.04
Tris(trimethylsilyl)phosphite ≥96%
Tris(trimethylsilyl)phosphite

Purity: 96% CAS No.: 1795-31-9 Molecular Formula: [(CH3)3SiO]3P Formula Weight: 298.54 Physical Form: Liquid Density: 0.893 Refractive Index: 1.409 Sensitivity: Air and Moisture Sensitive Boiling Point: 80°/8mm MDL No.: MFCD00015588

Formula: C₉H₂₇O₃PSi₃
Pto. Ebullición: 80 °C (8 mmHg)
Densidad: 0,893
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00015588
Núm. CAS: 1795-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR