VWR® VP 220, Chemical Duty, High Performance Diaphragm Pump
Proveedor: VWR Collection
181-0310EA 2960 EUR
181-0310
VWR® VP 220, Chemical Duty, High Performance Diaphragm Pump
Bombas Bombas de diafragma
The VP 220 is ideal for rotary evaporators providing the powerful vacuum needed for demanding evaporations without the use of oil pumps. VP 220 has built-in Advanced Vapour Management (AVM) which means precise control capability for the distillation of organic solvents and applications handling aggressive vapours and gases. The VP 220 is especially effective for rotary evaporation of low boiling point solvents such as acetone and ethanol and is able to rapidly strip DMF at 35 °C. The pump parts are all made of corrosion resistant materials such as PTFE and Kalrez to handle aggressive vapours,More Product Information