Bombas, de membrana, LABOPORT®
Proveedor: KNF
N035AN.18 N86KN.18 N86KT.18 N816.3KT.18 N816.1.2KT.18 N035.3AT18IP20 N816.3KT.45.18
181-3233EA 870 EUR
181-3233 181-3281 181-3282 181-3135 181-3109 181-0344 181-3917
Bombas, de membrana, LABOPORT®
Bombas Bombas de diafragma
Single- or dual-headed, dry running diaphragm pumps for a wide variety of applications in laboratories. They transfer and evacuate without affecting the media, i.e. media remains uncontaminated. The pump includes a PTFE-coated diaphragm with patented tension optimisation, which allows the pump to be smaller and prolongs the service life of the diaphragm, it is ideal for aggressive/corrosive gases and vapors.
  • 100% oil-free transfer, evacuation and compression, so media remains uncontaminated
  • Version available for neutral to mildly aggressive gases and