Aminophenylmorpholinone, Pharmaceutical Analytical Impurities

Proveedor: USP

1A01530
1A01530.EA 1050 EUR
1A01530.
Aminophenylmorpholinone, Pharmaceutical Analytical Impurities
Normativas Estándares orgánicos

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Order Now

ESPECIFICACIONES

Descripción Aminophenylmorpholinone
Synonym(s) 4-(4-aminophenyl)morpholin-3-one
Environmentally Preferable
CAS No. 438056-69-0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR