Anti-CD3E Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: 145-2C11]
Proveedor: Tonbo Biosciences
20-0031-U100 20-0031-U025
20-0031-U100EA 104 EUR
20-0031-U100 20-0031-U025
Anti-CD3E Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: 145-2C11]
Anticuerpos
The 145-2C11 antibody is specific for mouse CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses includingMore Product Information