Anti-CD45RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]

Proveedor: Tonbo Biosciences
20-0458-T025 20-0458-T100
20-0458-T025EA 51.2 EUR
20-0458-T025 20-0458-T100
Anti-CD45RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]
Anticuerpos
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, includeMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno PTPRC
Environmentally Preferable
Nombre del antígeno Protein tyrosine phosphatase receptor type C
Sinónimos de antígenos CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado APC (Allophycocyanin)
Clon HI100
Reactividad Chimpanzee,Human
Huésped Mouse
ID del gen 5788
Isotipo IgG2b kappa
Western blot Yes
InmunoQuímica Yes
Citometría de flujo Yes
Neutralización Yes
Adsorción cruzada none
Temperatura de almacenamiento Cool
Shipping temperature Ice
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR