Anti-IL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: JES6-5H4]
Proveedor: Tonbo Biosciences
20-7021-U025 20-7021-U100
20-7021-U025EA 61.8 EUR
20-7021-U025 20-7021-U100
Anti-IL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: JES6-5H4]
Anticuerpos
The JES6-5H4 antibody binds to mouse Interleukin-2 (IL-2), a 17 kDa cytokine that is secreted by activated T cells. This cytokine acts as an autocrine stimulatory factor for T cell proliferation, differentiation and survival, as well as being an effective activator of B cells and inducer of NK cell cytotoxic functions. IL-2 plays a key modulatory role in the differentiation of T cells toward either Th17 or T regulatory (Treg) cell types and is important for Treg cell function. The IL-2 receptor (IL-2R) is a complex consisting of an IL-2R alpha chain (CD25), along with the IL-2R beta chain (More Product Information