Bovine fibrinogen

Proveedor: SEQENS

2000-55-100ML 2000-55-500ML 2000-55-1L
2000-55-100MLEA 88 EUR
2000-55-100ML 2000-55-500ML 2000-55-1L
Bovine fibrinogen
Proteínas y Péptidos

This ready-to-use 5% frozen solution enables a direct use of the product without further manipulation.


  • Total proteins: >35,0 g/L
  • Shelf life: 3 years
  • Sterile filtered 0,2 µm


SEQENS IVD provides highly purified fibrinogen manufactured according to Cohn fractionation method. The plasma used is collected from healthy animals of French origin. Bovine Fibrinogen is developed for RPF supplement, used with Baird-Parker agar, for the detection of Staphylococcus aureus.


Full traceability and batch-to-batch repeatability guaranteed.


Certificaciones: Bovine plasma used for the manufacture of this product is collected from French healthy cattle declared free from BSE disease in its clinical forms. It is sourced in EU-approved French slaughterhouses.


Información de suministro: Supplied as a ready to use solution in aqueous saline buffer containing trisodium citrate and sodium chloride.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR