Acetona ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS para análisis

Proveedor: VWR Chemicals
Peligro

Sinónimos: Propanona

20066.310EA 33.4 EUR
20066.310 20066.365 20066.423 20066.467 20066.558 20066.296 20066.321 20066.330 20066.930
Acetona ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS para análisis
Acetona
Formula: CH₃COCH₃
Pto. Ebullición: 56,2 °C (1013 hPa)
Pto. de fusión: –95,4 °C
Densidad: 0,792 g/cm³ (20 °C)
Pto. de inflamación: <–20 °C (copa cerrada)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008765
Núm. CAS: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
IR: 01-2119471330-49
Índice Merck: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay (on anhydrous substance) Min. 99.8 %
Appearance of solution Passes test Ph.Eur.
Aqueous solution Passes test
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
Insolubility in water Passes test
IR Spectrum Passes test
Related substances Passes test Ph.Eur.
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0003 meq/g
Boiling point 56.0 - 56.7 °C
Colouration Max. 10 APHA
Density (20/4) 0.790 - 0.792
Density (20/20) 0.790 - 0.793
Aldehydes (as HCHO) Max. 10 ppm
Ethanol Max. 100 ppm
Evaporation residue Max. 5 ppm
Methanol Max. 0.05 %
Propan-2-ol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 2 ppm
Water Max. 0.2 %
PO4 (Phosphate) Max. 100 ppb
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR