Diamond pen
Proveedor: USBECK Laborgeräte
5240 5241
201-0391EA 39.5 EUR
201-0391 201-0392
Diamond pen
Bolígrafos Bolígrafos de tinta
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.