Centrifuge tubes, glass, Quickfit®
Proveedor: SciLabware
BC24/C14T
201-8267EA 248 EUR
201-8267
Centrifuge tubes, glass, Quickfit®
Tubos Tubos de centrífuga
Borosilicate glass, conical bottom, with Quickfit® socket.
  • Precision-ground Quickfit® socket to accept tapered stopper
  • Each tube is marked with its weight to the nearest 0,1 g to enable tubes of similar weight to be selected from any batch
  • Can be centrifuged at rcf values up to 3000×g
Joint size: 14/15