Anti-IgG2a Goat Polyclonal Antibody (CF™ 680)

Proveedor: Biotium
20263 20263-1
20263.EA 419 EUR
20263. 20263-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 680)
Anticuerpos
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR