Anti-IgG Goat Polyclonal Antibody (CF®583R)
Proveedor: Biotium
20904-50UL 20904-500UL
20904-50ULEA 103 EUR
20904-50UL 20904-500UL
Anti-IgG Goat Polyclonal Antibody (CF®583R)
Anticuerpos
This is a highly cross-adsorbed goat anti-mouse IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, horse, human, rabbit, and swine serum. This antibody is not cross-adsorbed against rat. For staining rat tissue, or co-staining with rat antibodies, we recommend using Goat Anti-Mouse IgG (H L), Highly Cross-Adsorbed (Min x Rat).
  • Highly-cross adsorbed for specific staining with minimal background
  • Available with 19 bright and photostable CF® dyes
  • Biotin and