Microtube racks, Handyrack series
Proveedor: Greiner Bio-One
999291_1 999293_1 999203_1 999207_1
GREI999291_1EA 4.6 EUR
GREI999291_1 212-0367 212-0369 212-0370
Microtube racks, Handyrack series
Gradillas Tube Racks
PP.
Racks available in an array of colours for easy identification.
Handyrack 80
Holds larger microtubes and skirted 0,5 ml microtubes.
Handyrack 96
Ideal for pipetting to and from 96-well plates.
Información para pedidos: Lids need to be ordered separately. Please enquire for details of lids and other colours of rack that are available.