Test tube racks, Switch-Grid, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F187450045 F187450011 F187450014
212-0691EA 53 EUR
212-0691 212-0687 212-0690
Test tube racks, Switch-Grid, SP Bel-Art
Gradillas Tube Racks
Polyoxymethylene, coloured.
  • Test tubes with 10 to 30 mm diameter fit in 5 colour coded grid inserts
  • Three tier rack holds tubes securely in place
  • Rubber feet keep racks from sliding on benchtop and also provide secure stacking while in storage
  • Can be cleaned with mild detergent, ethanol or isopropyl alcohol
The innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mildMore Product Information