Test tubes racks, wire
Proveedor: HEATHROW
HS23108 HS23072
212-3492EA 34.5 EUR
212-3492 212-3491
Test tubes racks, wire
Gradillas Tube Racks
Tube racks, made of chemically-resistant epoxy-coated steel wire.Durable racks for transport or sample handling.
  • With slotted bottom
  • Holds either 13 and 16 mm tubes
  • Very good chemical resistance, autoclavable