Beakers, low form, Nalgene®
Proveedor: Thermo Fisher Scientific
1201-0030 1201-0050 1201-0100 1201-0150 1201-0250 1201-0400 1201-0600 1201-1000 1201-2000 1201-4000
213-7100EA 44.9 EUR
213-7100 213-7101 213-7102 213-7103 213-7104 213-7105 213-7106 213-7107 213-7108 213-7109
Beakers, low form, Nalgene®
Vasos de precipitados
PPCO, translucent.
  • Very good chemical resistance
  • Heat resistant up to +121 °C, autoclavable at +121 °C (max. 20 minutes)
  • Smooth surfaces for easy, fast cleaning
Designed for general laboratory use. With spout and easy to read printed graduations with size code, plastic code, maximum temperature for use and 'Not flameproof' symbol. Ribbed for easy stacking. No drip, single spout design provides precision pouring control.
ISO 7056
Certificaciones: ISO 7056