Flasks acc. to Kjeldahl with standard ground joint
Proveedor: SciLabware
FK500/3L FK800/3L FK300/3L
212-0592EA 41.9 EUR
212-0592 214-0420 BIBBFK300/3L
Flasks acc. to Kjeldahl with standard ground joint
Matraces Matraces Kjeldahl
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For the determination of nitrogen by the Kjeldahl method. Extra long necks act as air condensers during digestion stage and reduce entrainment during distillation stage.